Manganese(II) sulphate monohydrate, powder USP
Supplier: Spectrum Chemical
Warning
Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate
M1114-12KG M1114-2.5KG M1114-50KGBL
SPCMM1114-12KGEA 1540 EUR
SPCMM1114-12KG SPCMM1114-2.5KG SPCMM1114-50KGBL
Manganese(II) sulphate monohydrate, powder USP
Manganese (II) sulphate monohydrate
Form: Powder
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763
Order Now
Specification Test Results
Assay 98% Min.