Magnesium stearate BP, JP, NF
Supplier: Spectrum Chemical
Warning
Synonyms: Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt
MA130-12KG MA130-2.5KG MA130-45KGBL
SPCMMA130-12KGEA 1660 EUR
SPCMMA130-12KG SPCMMA130-2.5KG SPCMMA130-45KGBL
Magnesium stearate BP, JP, NF
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
Melting Pt: 130…140 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 13,05714
Order Now
Specification Test Results
Assay 4% Min.