Potassium benzoate NF
Supplier: Spectrum Chemical
Synonyms: Benzoic acid potassium salt
P1181-12KG P1181-2.5KG P1181-45KGBL
SPCMP1181-12KGEA 831 EUR
SPCMP1181-12KG SPCMP1181-2.5KG SPCMP1181-45KGBL
Potassium benzoate NF
Potassium benzoate
Formula: C₇H₅KO₂
MW: 160.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013061
CAS Number: 582-25-2
EINECS: 209-481-3
Order Now
Specification Test Results
Assay 99% Min.