L-Phenylalanine USP
Supplier: Spectrum Chemical
Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
P1582-1KG P1582-50KGBL P1582-5KGBL
SPCMP1582-1KGEA 3260 EUR
SPCMP1582-1KG SPCMP1582-50KGBL SPCMP1582-5KGBL
L-Phenylalanine USP
L-α-Phenylalanine
Formula: C₉H₁₁NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271
Order Now
Specification Test Results
Assay 98.5% Min.