Polysorbate 60 NF
Supplier: Spectrum Chemical
Synonyms: Sorbitan monostearate, ethoxylated, Polyoxyethylene sorbitan monostearate
PO136-200LTBL PO136-20LT PO136-4LTGL
SPCMPO136-200LTBLEA 8100 EUR
SPCMPO136-200LTBL SPCMPO136-20LT SPCMPO136-4LTGL
Polysorbate 60 NF
Polysorbate 60
Formula: C₆₄H₁₂₆O₂₆
Storage Temperature: Ambient
MDL Number: MFCD01780110
CAS Number: 9005-67-8
EINECS: 500-020-4