2,6-Di-tert-butyl-p-cresol, granular NF
Supplier: Spectrum Chemical
Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
B1196-12KGBL B1196-2.5KG B1196-500GM
SPCMB1196-12KGBLEA 2570 EUR
SPCMB1196-12KGBL SPCMB1196-2.5KG SPCMB1196-500GM
2,6-Di-tert-butyl-p-cresol, granular NF
2,6-Di-tert-butyl-p-cresol
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₁₅H₂₄O
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 69…70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00011644
CAS Number: 128-37-0
EINECS: 204-881-4
Merck Index: 13,01547
Order Now
Specification Test Results
Assay 99% Min.