Sodium acetate trihydrate, granular USP
Supplier: Spectrum Chemical
SO105-100KGBL SO105-12KG SO105-2.5KG SO105-50KGBL
SPCMSO105-100KGBLEA 3050 EUR
SPCMSO105-100KGBL SPCMSO105-12KG SPCMSO105-2.5KG SPCMSO105-50KGBL
Sodium acetate trihydrate, granular USP
Sodium acetate trihydrate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99% Min.