Sodium salicylate, crystals USP
Supplier: Spectrum Chemical
Warning
Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt
SO205-12KG SO205-2.5KG SO205-50KGBL
SPCMSO205-12KGEA 1670 EUR
SPCMSO205-12KG SPCMSO205-2.5KG SPCMSO205-50KGBL
Sodium salicylate, crystals USP
Sodium salicylate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819
Order Now
Specification Test Results
Assay 99.5% Min.