Sorbitan monostearate NF
Supplier: Spectrum Chemical
Synonyms: Sorbitan monooctadecanoate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan stearate
SPA63-12KG SPA63-2.5KG SPA63-90KGBL
SPCMSPA63-12KGEA 1600 EUR
SPCMSPA63-12KG SPCMSPA63-2.5KG SPCMSPA63-90KGBL
Sorbitan monostearate NF
Sorbitan monostearate
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₂₄H₄₆O₆
MW: 430.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9
Order Now
Specification Test Results
Assay 68% Min.