(±)-1,3-Butanediol
Supplier: Spectrum Chemical
Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane
B2147-200LTBL B2147-20LT
SPCMB2147-200LTBLEA 4800 EUR
SPCMB2147-200LTBL SPCMB2147-20LT
(±)-1,3-Butanediol
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 206…208 °C (1013 hPa)
Melting Pt: < –50 °C
Density: 1.004…1.006 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566
Order Now
Specification Test Results
Assay 99% Min.