Benzoic acid, crystals USP, Ph. Eur., BP, JP
Supplier: Spectrum Chemical
Warning
BE140-12KG BE140-2.5KG BE140-25KGBL
SPCMBE140-12KGEA 1530 EUR
SPCMBE140-12KG SPCMBE140-2.5KG SPCMBE140-25KGBL
Benzoic acid, crystals USP, Ph. Eur., BP, JP
Benzoic acid
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.5% Min.