L(-)-Tyrosine USP
Supplier: Spectrum Chemical
Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
T1153-25KGBL T1153-50KGBL T1153-5KG T1153-100GM
SPCMT1153-25KGBLEA 0 EUR
SPCMT1153-25KGBL SPCMT1153-50KGBL SPCMT1153-5KG SPCMT1153-100GM
L(-)-Tyrosine USP
L(-)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907
Order Now
Specification Test Results
Assay 98.5% Min.