Calcium sulphate dihydrate NF
Supplier: Spectrum Chemical
Synonyms: Plaster of Paris
CA170-12KG CA170-2.5KG CA170-45KGBL
SPCMCA170-12KGEA 1020 EUR
SPCMCA170-12KG SPCMCA170-2.5KG SPCMCA170-45KGBL
Calcium sulphate dihydrate NF
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711
Order Now
Specification Test Results
Assay 98% Min.