2-Nitro-5-thiocyanatobenzoic acid, crystalline powder
Supplier: MP Biomedicals
Synonyms: 2-Nitro-5-Thiocyanobenzoic acid
0215592991 0215592983
ICNA0215592991EA 375 EUR
ICNA0215592991 ICNA0215592983
2-Nitro-5-thiocyanatobenzoic acid, crystalline powder
2-Nitro-5-thiocyanatobenzoic acid
2-nitro-5-thiocyanatobenzoic acid (NTCB) is commonly used to cyanylate and cleave proteins at cysteine residues.
Formula: C₈H₄N₂O₄S
MW: 224.2 g/mol
MDL Number: MFCD00007139
CAS Number: 30211-77-9