VWR® Traceable®, USB Data Logging Refrigerator/Freezer Thermometers
Supplier: VWR Collection
620-2442EA 428 EUR
620-2442 620-2443
VWR® Traceable®, USB Data Logging Refrigerator/Freezer Thermometers
Thermometers Digital Thermometers
Data logging thermometer with user-defined time intervals. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators.
  • User-defined data logging interval of 1 observation/minute to 1 observation/24 hours
  • Rolling memory structure, maintains most recent 1 000 000 temperature observations
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Stores up to 10 unique alarm events
  • Hassle-free retrieval of data - unit can remain in use while downloading and analysing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Recorded data (CSV file) to be transferred from thermometer to PC or Mac using a USB flash drive (not included)
  • High impact, chemical resistant ABS plastic case
Certifikace: CE marked. Meets current CDC requirements for vaccine storage and monitoring. Certified and Traceable® to NIST.
Delivery information: Delivered with platinum RTD sensors, stainless steel 316 probe(s), stand, VELCRO®, magnetic strips, wall mount, Traceable® certificate and batteries.