Cold resistant gloves, uvex unilite thermo
Supplier: UVEX
6059310 6059311 6059307 6059308 6059309
112-4534EA 93.7 EUR
112-4534 112-4535 112-4531 112-4532 112-4533
Cold resistant gloves, uvex unilite thermo
Gloves
These winter safety gloves feature a dual-layer construction: The inner glove is made of acrylic and a new wool mix, the outer glove is made of polyamide/elastane. The gloves have a flexible polymer coating on palm and fingertips.
  • Extremely robust coating is flexible even at low temperatures
  • Double layer construction ensures effective insulation
  • Excellent dexterity, high flexibility
  • Mechanical strength
Ideal for forklift truck drivers and for use in refrigerated warehouses or when working in cold conditions. For use in dry and slightly moist areas.
Certifikace: EN 388:2016 (3131X), EN 511 (010)
Food contact approved