Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
LCK114 LCK614 LCK914 LCK014 LCK314 LCK514 LCK1014 LCK1414
705-3033EA 134 EUR
705-3033 705-3037 705-3038 705-3032 705-3034 705-3036 705-3075 705-3073
Test kits, photometric, COD, LCK cuvette tests
Environmental Test Kits
Cuvette tests for water testing applications. Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient, error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour coding
Storage conditions: 15 to 25 °C (protect from light)
Certifikace: ISO 6060-1989, DIN 38409-H41-H44
Informace o príslušenství: Cuvette tests are officially approved for many legally required consent limits and with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.