Aristolochic acid mixture of I and II 96%
Supplier: Thermo Scientific
|
Danger
|
ACRO226095000EA
580
EUR
ACRO226095000
ACRO226091000
Aristolochic acid mixture of I and II 96%
Aristolochic acid
Formula:
C₁₇H₁₁NO₇ MW: 341,275 g/mol Melting Pt: 269…270 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004996 CAS Number: 313-67-7 UN: 1544 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % (typically I + II isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...