cis-2-Aminomethyl-1-cyclohexanol hydrochloride 99%
Supplier: Thermo Fisher Scientific
Warning
265880010 265882500
ACRO265880010EA 289 EUR
ACRO265880010 ACRO265882500
cis-2-Aminomethyl-1-cyclohexanol hydrochloride 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride
Appearance: White Crystalline powder
Formula: C₇H₁₅NO·HCl
MW: 165.66 g/mol
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00143984
CAS Number: 24947-68-0
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %