(S)-(+)-Camptothecin 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
276721000 276720010
ACRO276721000EA 228 EUR
ACRO276721000 ACRO276720010
(S)-(+)-Camptothecin 98%
(S)-(+)-Camptothecin
CAS No.: 7689-03-4
Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
HPLC ≥97.5 %