4-Cyanostyrene 95% Stabilised
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Ethenylbenzonitrile
292910025 292910100
ACRO292910025EA 120 EUR
ACRO292910025 ACRO292910100
4-Cyanostyrene 95% Stabilised
4-Cyanostyrene
CAS No.: 3435-51-6
Formula: C₉H₇N
MW: 129.162 g/mol
Boiling Pt: 56…58 °C (10 mmHg)
Storage Temperature: Freezer
MDL Number: MFCD00080445
CAS Number: 3435-51-6
UN: 3276
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5765 to 1.5785 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.05 % tert.butylcatechol