1-Methylimidazolidin-2-one 98+%
Supplier: Thermo Scientific
Warning
Synonyms: 1-Methyl-2-imidazolidinone
304155000 304150020
ACRO304155000EA 68.1 EUR
ACRO304155000 ACRO304150020
1-Methylimidazolidin-2-one 98+%
1-Methylimidazolidin-2-one
Appearance: Off-white to beige Crystalline solid
Formula: C₄H₈N₂O
MW: 100.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800601
CAS Number: 694-32-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Off-white to beige Crystalline solid
Infrared spectrum Conforms
GC ≥98.0 %