Fumonisin B1 96%
Supplier: Thermo Scientific
Warning
328250010 328250050
ACRO328250010EA 178 EUR
ACRO328250010 ACRO328250050
Fumonisin B1 96%
Fumonisin B1
CAS No.: 116355-83-0
Formula: C₃₄H₅₉NO₁₅
MW: 721.838 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133349
CAS Number: 116355-83-0
Order Now
Specification Test Results
Appearance White to tan Powder or granules or solid
HPLC ≥95 %