Nonylphenol (mixture of isomers) 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
416240010
416240050
ACRO416240010EA
153
EUR
ACRO416240010
ACRO416240050
Nonylphenol (mixture of isomers) 99%
Nonylphenol (mixture of isomers)
Formula:
C₁₅H₂₄O Flash Pt: 141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002396 CAS Number: 84852-15-3 UN: 3145 ADR: 8,II |
Specification Test Results
Appearance | Clear viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5105 to 1.5165 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥98.5 % |
Color scale | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...