Phenyl-p-benzoquinone 95%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Phenyl-1,4-benzoquinone, [1,1'-Biphenyl]-2,5-dione
417380250 417381000
ACRO417380250EA 120 EUR
ACRO417380250 ACRO417381000
Phenyl-p-benzoquinone 95%
Phenyl-p-benzoquinone
CAS No.: 363-03-1
Formula: C₁₂H₈O₂
MW: 184.194 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00001599
CAS Number: 363-03-1
Order Now
Specification Test Results
Appearance Yellow-brown Powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥94 %