DL-α-Tocopherol acetate 98%
Supplier: Thermo Fisher Scientific
Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
421070010 421070050
ACRO421070010EA 28.1 EUR
ACRO421070010 ACRO421070050
DL-α-Tocopherol acetate 98%
DL-α-Tocopherol acetate
Flash Point: >110°C
Appearance: Clear light yellow to light green Viscous liquid
Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
Flash Pt: 257 °C
Storage Temperature: Ambient
MDL Number: MFCD00072042
CAS Number: 7695-91-2
Order Now
Specification Test Results
Appearance Clear light yellow to light green Viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥97.5 %