Petroleum spirit, 35…60 °C ACS
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Gasoline, Petroleum ether, Petrol, Benzine
422995000 422990050
ACRO422995000EA 26.7 EUR
ACRO422995000 ACRO422990050
Petroleum spirit, 35…60 °C ACS
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear liquid
Boiling point 35 to 60°C
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity Passes test