Benzylhydrazine dihydrochloride 97%

Supplier: Thermo Scientific
Warning

105960050 105960250 105961000
ACRO105960050EA 51.1 EUR
ACRO105960050 ACRO105960250 ACRO105961000
Benzylhydrazine dihydrochloride 97%
Benzylhydrazine dihydrochloride

CAS No.: 20570-96-1

Formula: C₇H₁₀N₂
MW: 195.09 g/mol
Melting Pt: 138…143 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012921
CAS Number: 20570-96-1

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 138 to 143 °C
Titration Argentometric ≥96.0 %
Color scale ≤100 APHA
Titration with NaOH ≥96.0 %
Impurity ≤3.0 % Tributyl phosphate
Acid value 259 to 267 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR