Rutin hydrate 97+%
Supplier: Thermo Scientific
132390050 132391000 132395000
ACRO132390050EA 39.6 EUR
ACRO132390050 ACRO132391000 ACRO132395000
Rutin hydrate 97+%
Rutin hydrate
CAS No.: 153-18-4
Formula: C₂₇H₃₀O₁₆
MW: 610.52 g/mol
Melting Pt: 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006830
CAS Number: 207671-50-9
Order Now
Specification Test Results
Appearance Yellow to yellow-green to straw yellow Powder
Infrared spectrum Conforms
Loss on drying 5.0 % to 9.0 % (500 mg, 105°C, 3 h)
Ash ≤0.5 %
Titration with NaOH 97.0 to 102.0 % (On dry basis)