trans-Crotonyl chloride 90% tech.
Supplier: Thermo Scientific
Danger
Synonyms: (2E)-but-2-enoyl chloride, (E)-2-Butenoyl chloride, trans-2-Butenoyl chloride
155910050 155911000 155915000
ACRO155910050EA 30.1 EUR
ACRO155910050 ACRO155911000 ACRO155915000
trans-Crotonyl chloride 90% tech.
trans-Crotonyl chloride
CAS No.: 625-35-4
Formula: C₄H₅ClO
MW: 104.533 g/mol
Boiling Pt: 120…123 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000734
CAS Number: 625-35-4
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4585 to 1.4615 (20°C, 589 nm)
GC ≥88 %
UV at 430 nm A: ≤0.01
Color scale ≤10 APHA