3-Amino-4-pyrazolecarbonitrile 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-Aminopyrazole-4-carbonitrile, 3-Amino-1H-pyrazole-4-carbonitrile, 3-Aminopyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole 3-Amino-4-pyrazolecarbonitrile 5-Amino-4-cyanopyrazole 3-Amino-1H-pyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole
166850010
166850050
166850250
ACRO166850010EA
23.4
EUR
ACRO166850010
ACRO166850050
ACRO166850250
3-Amino-4-pyrazolecarbonitrile 98%
3-Amino-4-pyrazolecarbonitrile
Appearance: Light yellow to orange-brown Crystalline powder
Formula:
C₄H₄N₄ MW: 108.10 g/mol Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005237 CAS Number: 16617-46-2 UN: 2811 ADR: 6.1 III |
Specification Test Results
Appearance | Light yellow to orange-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 172 to 176 °C |
Titration with HClO4 | ≥97.5 % |
Acid value | ≤2.0 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...