2-Sulphobenzaldehyde sodium salt 90% tech.
Supplier: Thermo Fisher Scientific
Synonyms: 2-Formylbenzenesulphonic acid sodium salt, Sodium 2-formylbenzenesulphonate, Sodium 2-Formylbenzenesulphonate
219325000 219320025
ACRO219325000EA 118 EUR
ACRO219325000 ACRO219320025
2-Sulphobenzaldehyde sodium salt 90% tech.
2-Sulphobenzaldehyde sodium salt
CAS No.: 1008-72-6
Formula: C₇H₆O₄S
MW: 208.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007478
CAS Number: 1008-72-6
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥88 %
Sodium chloride (NaCl) ≤3 %