Potassium-tert-butoxide 20% (w/w) in THF, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide
364990500 364992500
ACRO364990500EA 121 EUR
ACRO364990500 ACRO364992500
Potassium-tert-butoxide 20% (w/w) in THF, pure
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Density: 0.929 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00012162
CAS Number: 865-47-4
UN: 2920
ADR: 8 I
Order Now
Specification Test Results
Appearance Colorless to amber Solution
Strength 19 to 21 % (Acidimetric)