(R)-(S)-Josiphos 97+%
Supplier: Thermo Scientific
Synonyms: (2R)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (2Sp)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, (R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine
370671000 370675000
ACRO370671000EA 69.9 EUR
ACRO370671000 ACRO370675000
(R)-(S)-Josiphos 97+%
(R)-(S)-Josiphos
CAS No.: 155806-35-2
Formula: C₃₆H₄₄FeP₂
MW: 594.541 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800284
CAS Number: 155806-35-2
Order Now
Specification Test Results
Appearance Orange Powder
Infrared spectrum Conforms
HPLC ≥97.0 %