Methylmagnesium bromide 3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
391121000 391128000
391121000.EA 102 EUR
391121000. 391128000.
Methylmagnesium bromide 3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Methylmagnesium bromide
Formula: CH₃BrMg
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000041
CAS Number: 75-16-1
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 34 to 36 %