(2-((Dimethylamino)methyl)phenyl)boronic acid 98%
Supplier: Thermo Scientific
|
|
ACRO359241000EA
120
EUR
ACRO359241000
(2-((Dimethylamino)methyl)phenyl)boronic acid 98%
(2-((Dimethylamino)methyl)phenyl)boronic acid
Formula:
C₉H₁₄BNO₂ MW: 179,0240 g/mol |
MDL Number:
MFCD01318999 CAS Number: 85107-53-5 |
Specification Test Results
Appearance | Beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...