Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Supplier: Thermo Fisher Scientific
360140500 360142500
ACRO360140500EA 190 EUR
ACRO360140500 ACRO360142500
Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00148339
CAS Number: 7631-86-9
Order Now
Specification Test Results
Appearance White to off-white Powder
Particle size 40 to 63 µm
Elemental Analysis ≥11.6 % (C)
Loading ≥1.21 mmol/g
Volatile matter ≤4 % (110°C)