(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%, 98% ee
Supplier: Thermo Fisher Scientific
Warning
362270010 362270050
ACRO362270010EA 530 EUR
ACRO362270010 ACRO362270050
(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%, 98% ee
(1R,3S)-3-Aminocyclopentanecarboxylic acid
Appearance: Off-white to beige Crystalline powder
Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
Melting Pt: 172.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00211289
CAS Number: 71830-08-5
Order Now
Specification Test Results
Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)