(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride 95%
Supplier: Thermo Scientific
Warning
Synonyms: (1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, (1R,2S,3R,5R)-3-Amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride
362520010 362522500 362520050
ACRO362520010EA 241 EUR
ACRO362520010 ACRO362522500 ACRO362520050
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride 95%
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride
CAS No.: 79200-57-0
Formula: C₆H₁₄ClNO₃
MW: 183.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06799068
CAS Number: 79200-57-0
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥94 %
Specific optical rotation -16° to -20° (20°C, 589 nm) (c=1, methanol)