1,1-Dimethylpropargylamine 95%
Supplier: Thermo Scientific
Danger
Synonyms: 2-Methylbut-3-yn-2-amine
175620010 175620050 175620250
ACRO175620010EA 22.2 EUR
ACRO175620010 ACRO175620050 ACRO175620250
1,1-Dimethylpropargylamine 95%
1,1-Dimethylpropargylamine
Appearance: Clear colorless to yellow Liquid
Formula: C₅H₉N
MW: 83.134 g/mol
Boiling Pt: 79…80 °C (760 mmHg)
Density: 0.79 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008052
CAS Number: 2978-58-7
UN: 2733
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4260 (20°C, 589 nm)
GC ≥94 %
HPLC ≥97.5 %