4-Methylpiperazine-1-ethanol 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-(2-Hydroxyethyl)-4-methylpiperazine
384830010 384830050 384832500
ACRO384830010EA 81.7 EUR
ACRO384830010 ACRO384830050 ACRO384832500
4-Methylpiperazine-1-ethanol 98%
4-Methylpiperazine-1-ethanol
Appearance: Light yellow Low melting solid
Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Boiling Pt: 54…55 °C (.5mbar)
Storage Temperature: Ambient
MDL Number: MFCD03840691
CAS Number: 5464-12-0
Order Now
Specification Test Results
Appearance Light yellow Low melting solid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4870 (20°C, 589 nm)
GC ≥97.5 %