3-Quinolinol 97+%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Hydroxyquinoline
391370010 391372500
ACRO391370010EA 412 EUR
ACRO391370010 ACRO391372500
3-Quinolinol 97+%
3-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00169018
CAS Number: 580-18-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 198 to 202 °C
HPLC ≥97.0 %