Hydrogen chloride ∼0.5 M in methanol
Supplier: Thermo Scientific
Danger
391741000 391748000
ACRO391741000EA 32.4 EUR
ACRO391741000 ACRO391748000
Hydrogen chloride ∼0.5 M in methanol
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Melting Pt: -30 °C
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 0.4 to 0.6 M