2-Bromo-6-formylpyridine 97%
Supplier: Thermo Scientific
Warning
Synonyms: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde
387420010 387420050
ACRO387420010EA 37.5 EUR
ACRO387420010 ACRO387420050
2-Bromo-6-formylpyridine 97%
2-Bromo-6-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2
Order Now
Specification Test Results
Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥96.0 %