5-Fluoro-2-nitrobenzoic acid 98%
Supplier: Thermo Scientific
Warning
398870010 398870050
ACRO398870010EA 24.7 EUR
ACRO398870010 ACRO398870050
5-Fluoro-2-nitrobenzoic acid 98%
5-Fluoro-2-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00055635
CAS Number: 320-98-9
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥97.5 %