Palladium(II) nitrate hydrate
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Palladium dinitrate hydrate, Palladous nitrate hydrate
389910010 389910050 389910250
ACRO389910010EA 156 EUR
ACRO389910010 ACRO389910050 ACRO389910250
Palladium(II) nitrate hydrate
Palladium(II) nitrate hydrate
CAS No.:
Formula: N₂O₆Pd·xH₂O
MW: 230.43 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149818
CAS Number: 207596-32-5
UN: 3085
ADR: 5.1 II
Order Now
Specification Test Results
Appearance Brown to brown-black Crystals
Palladium (Pd) 39 to 46 %