2-Bromo-3-formylpyridine 96%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde
399670010 399670050
ACRO399670010EA 37.6 EUR
ACRO399670010 ACRO399670050
2-Bromo-3-formylpyridine 96%
2-Bromo-3-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Boiling Pt: 100 °C (3 mmHg)
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD04966945
CAS Number: 128071-75-0
Order Now
Specification Test Results
Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 75 °C
HPLC ≥95.0 %
NMR (1H-NMR) Consistent with structure