Histamine (free base) 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine, Histamine
411710010 411710050 411710250
ACRO411710010EA 76.4 EUR
ACRO411710010 ACRO411710050 ACRO411710250
Histamine (free base) 97%
Histamine (free base)
Formula: C₅H₉N₃
MW: 111.15 g/mol
Boiling Pt: 167 °C (.8 mmHg)
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005210
CAS Number: 51-45-6
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %