Ethylaluminium dichloride 1.8 M in toluene, AcroSeal®
Supplier: Thermo Scientific
Danger
428041000 428048000
428041000.EA 83.8 EUR
428041000. 428048000.
Ethylaluminium dichloride 1.8 M in toluene, AcroSeal®
Ethylaluminium dichloride
Formula: C₂H₅AlCl₂
MW: 126.95 g/mol
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000457
CAS Number: 563-43-9
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
Strength 23 to 27 %