Triethyl borate 97%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
428651000 428658000
428651000.EA 45.5 EUR
428651000. 428658000.
Triethyl borate 97%, AcroSeal®
Triethyl borate
Formula: C₆H₁₅BO₃
MW: 145.99 g/mol
Boiling Pt: 117
Melting Pt: –84.5 °C
Density: 0.858 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009073
CAS Number: 150-46-9
UN: 1176
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3705 to 1.3775 (20°C, 589 nm)
GC ≥96.0 %